Skip to Content
Merck
All Photos(1)

Documents

M4943

Sigma-Aldrich

Anti-phospho-MEK4 (pSer257/pThr261) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, frog, mouse

technique(s)

western blot: 0.1-1.0 μg/mL using extracts prepared from GST-tagged fusion protein expressing MEK 4.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAP2K4(6416)
mouse ... Map2k4(26398)

General description

Mitogen-Activated Protein Kinase Kinase 4 (MEK 4 or MKK4) is a 42 kDa member of a tyrosine/threonine protein kinase family that activates the c-Jun NH2-terminal kinases (JNK), which is part of the inflammation/stress signaling pathway. The mutation of the MEK 4 gene in some carcinomas indicates that it may also have a role as a tumor suppressor. Phosphorylation of MEK4 by MEKK1 on serine 257 and threonine 261 in the catalytic domain activates the protein and enables MEK 4 to phosphorylate JNK
This antibody specifically recognizes human MEK 4 double phosphorylated at serine 257 and threonine 261. Mouse and frog MEK 4 (100% homology) may cross react.

Immunogen

synthetic MEK4-derived phosphopeptide that is phosphorylated at serine 257 and threonine 261.

Application

Anti-phospho-MEK4 (Tyr397) antibody may be used for detection by immunoblotting at a working concentration of 0.1-1.0 μg/mL.

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3, with 1.0 mg/mL BSA (IgG and protease free) and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Davis
Biochemical Society symposium, 64, 1-12 (1999-04-20)
The c-Jun N-terminal kinase (JNK) group of mitogen-activated protein kinases (MAP kinases) is activated by exposure of cells to environmental stress and by the treatment of cells with cytokines. The mechanism of activation of JNK is mediated by dual phosphorylation
A Cuenda
The international journal of biochemistry & cell biology, 32(6), 581-587 (2000-04-28)
The Mitogen-Activated Protein Kinase Kinase 4 (MKK4), a member of the MAP kinase kinase family, directly phosphorylates and activates the c-Jun NH2-terminal kinases (JNK), in response to cellular stresses and proinflammatory cytokines. JNK is a member of the MAP kinase
C Hagemann et al.
Cellular signalling, 13(12), 863-875 (2001-12-01)
MEK kinases (MEKKs) comprise a family of related serine-threonine protein kinases that regulate mitogen-activated protein kinase (MAPK) signalling pathways leading to c-Jun NH2-terminal kinase (JNK) and p38 activation, induced by cellular stress (e.g., UV and gamma irradiation, osmotic stress, heat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service