Skip to Content
Merck
All Photos(1)

Key Documents

LUC30059-1

Sigma-Aldrich

CloneID 1X COLONY PCR MASTER MIX

sufficient for 250 reactions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
Pricing and availability is not currently available.

usage

sufficient for 250 reactions

availability

available only in (India)

shipped in

dry ice

Looking for similar products? Visit Product Comparison Guide

General description

CloneID 1X Colony PCR Master Mix is simple and reliable solution for Colony PCR. Simply add your primers and a small amount of a colony from a plate, then cycle the reaction. No need to add water, optimize Magnesium concentration, or lyse cells. This master mix is robust under challenging conditions, such as genotyping directly from cells, so you get the most reproducible and reliable results every time. The product is also formulated with green gel loading dye for easy gel loading and visualization. It′s the fastest and easiest Colony PCR product available.

Legal Information

CloneID is a trademark of Lucigen Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service