Skip to Content
Merck
All Photos(1)

Key Documents

L7020

Sigma-Aldrich

Calcium levulinate dihydrate

Synonym(s):

γ-Ketovaleric acid calcium salt, 4-Oxopentanoic acid calcium salt, Levulinic acid calcium salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CH2COO · 1/2Ca · H2O
CAS Number:
Molecular Weight:
153.16
Beilstein:
3764203
EC Number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

O.CC(=O)CCC(=O)O[Ca]OC(=O)CCC(C)=O

InChI

1S/2C5H8O3.Ca.H2O/c2*1-4(6)2-3-5(7)8;;/h2*2-3H2,1H3,(H,7,8);;1H2/q;;+2;/p-2

InChI key

VPHDLUWYYKTYFR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service