Skip to Content
Merck
All Photos(1)

Key Documents

L4879

Sigma-Aldrich

Lumiflavine

powder

Synonym(s):

7,8,10-Trimethylbenzo[g]pteridine-2,4(3H,10H)-dione, 7,8,10-Trimethylisoalloxazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12N4O2
CAS Number:
Molecular Weight:
256.26
Beilstein:
269756
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

powder

storage temp.

−20°C

SMILES string

CN1c2cc(C)c(C)cc2N=C3C(=O)NC(=O)N=C13

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Lumiflavine (Lumiflavin) is produced by the photolysis of vitamin B2 (Riboflavin). Lumiflavine, a riboflavin uptake inhibitor, is used to study riboflavin uptake in intestinal epithelial (Caco-2) and other epithelial cells.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valeria Bafunno et al.
The Journal of biological chemistry, 279(1), 95-102 (2003-10-14)
We have studied the functional steps by which Saccharomyces cerevisiae mitochondria can synthesize FAD from cytosolic riboflavin (Rf). Riboflavin uptake into mitochondria took place via a mechanism that is consistent with the existence of (at least two) carrier systems. FAD
Vincent Sichula et al.
The journal of physical chemistry. B, 114(29), 9452-9461 (2010-07-06)
We investigated the oxidation behavior of 5-ethyl-4a-hydroxy-3-methyl-4a,5-dihydrolumiflavin (pseudobase Et-FlOH) in acetonitrile with the aim of determining if the two-electron oxidized Et-FlOH(2+) undergoes a release of hydroxyl cation and the production of 5-ethyl-3methyllumiflavinium cation (Et-Fl(+)). The focus of this work is
Hamid M Said et al.
The Journal of physiology, 566(Pt 2), 369-377 (2005-05-10)
In mammalian cells (including those of the ocular system), the water-soluble vitamin B2 (riboflavin, RF) assumes an essential role in a variety of metabolic reactions and is critical for normal cellular functions, growth and development. Cells of the human retinal
N B Sankaran et al.
The journal of physical chemistry. B, 113(5), 1522-1529 (2009-01-30)
The binding behavior of lumiflavin, a biologically vital ligand, with DNA duplexes containing an abasic (AP) site and various target nucleobases opposite the AP site is studied. Lumiflavin binds selectively to thymine (T) opposite the AP site in a DNA
Maxime T A Alexandre et al.
Biophysical journal, 97(1), 227-237 (2009-07-08)
Phototropins, major blue-light receptors in plants, are sensitive to blue light through a pair of flavin mononucleotide (FMN)-binding light oxygen and voltage (LOV) domains, LOV1 and LOV2. LOV2 undergoes a photocycle involving light-driven covalent adduct formation between a conserved cysteine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service