Skip to Content
Merck
All Photos(1)

Key Documents

L3669

Sigma-Aldrich

LPK-26

≥98% (HPLC), powder

Synonym(s):

2-(3,4-Dichlorophenyl)-N-methyl-N-[(1S)-1-(2-isopropyl)-2-(1-(3-pyrrolinyl))ethyl]acetamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24Cl2N2O · HCl
CAS Number:
Molecular Weight:
391.76
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

off-white

solubility

H2O: >10 mg/mL

storage temp.

2-8°C

SMILES string

Cl.CC(C)[C@@H](CN1CC=CC1)N(C)C(=O)Cc2ccc(Cl)c(Cl)c2

InChI

1S/C18H24Cl2N2O.ClH/c1-13(2)17(12-22-8-4-5-9-22)21(3)18(23)11-14-6-7-15(19)16(20)10-14;/h4-7,10,13,17H,8-9,11-12H2,1-3H3;1H/t17-;/m1./s1

InChI key

WOOCHAFXTYNYQE-UNTBIKODSA-N

Biochem/physiol Actions

LPK-26 is a potent, selective kappa opioid receptor agonist and an antinociceptive, devoid physical dependence properties.
LPK-26 is an analog of ICI-199441 and (-)U50,488H (classic kappa agonists). It has extremely high kappa affinity (0.64 nM) and low mu and delta receptor affnities (1170 and 10,000nM). LPK-26 produces an antinocicpetive effect with much higher potency in vivo than morphine or U50488. LPK-26 does not produce physical dependence in mice, but it does suppress naloxone-induced behavioral effects in mice treated with morphine.

Features and Benefits

This compound is featured on the Opioid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to opioid receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service