Skip to Content
Merck
All Photos(1)

Documents

I5759

SAFC

Methyl isobutyrate

Synonym(s):

Isobutyric acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200

density

0.89 g/mL at 25 °C (lit.)

storage temp.

2-8°C

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service