Skip to Content
Merck
All Photos(2)

Key Documents

I2127

Sigma-Aldrich

Isocytosine

≥99%

Synonym(s):

2-Amino-4-hydroxypyrimidine, 2-Aminouracil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5N3O
CAS Number:
Molecular Weight:
111.10
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

biological source

synthetic (organic)

Assay

≥99%

form

powder

solubility

acetic acid: 50 mg/mL, clear, colorless

SMILES string

NC1=NC=CC(=O)N1

InChI

1S/C4H5N3O/c5-4-6-2-1-3(8)7-4/h1-2H,(H3,5,6,7,8)

InChI key

XQCZBXHVTFVIFE-UHFFFAOYSA-N

Application

Isocytosine (2-aminouracil) is an isomer of cytosine used in physical chemical studies involving metal complex binding, hydrogen-bonding, and tautomerism and proton transfer effects in nucleobases.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X L Yang et al.
Biophysical journal, 75(3), 1163-1171 (1998-09-03)
Isoguanine (2-hydroxyladenine) is a product of oxidative damage to DNA and has been shown to cause mutation. It is also a potent inducer of parallel-stranded DNA duplex structure. The structure of the parallel-stranded DNA duplex (PS-duplex) 5'-d(TiGiCAiCiGiGAiCT) + 5'-d(ACGTGCCTGA), containing
A Michael Sismour et al.
Nucleic acids research, 33(17), 5640-5646 (2005-09-30)
Synthetic biology based on a six-letter genetic alphabet that includes the two non-standard nucleobases isoguanine (isoG) and isocytosine (isoC), as well as the standard A, T, G and C, is known to suffer as a consequence of a minor tautomeric
K J LaChance-Galang et al.
Inorganic chemistry, 40(3), 485-492 (2001-02-24)
Pentaammineruthenium moves on ambidentate nitrogen heterocycles by both rotation and linkage isomerization, which may affect the biological activity of potential ruthenium metallopharmaceuticals. The rapid rotation rates of [(NH3)5RuIII] coordinated to the exocyclic nitrogens of isocytosine (ICyt) and 6-methylisocytosine (6MeICyt) have
Deepali Gupta et al.
Inorganic chemistry, 43(11), 3386-3393 (2004-05-25)
Isocytosine (ICH) exists in solution as two major tautomers, the keto form with N1 carrying a proton (1a) and the keto form with N3 being protonated (1b). In water, 1a and 1b exist in equilibrium with almost equal amounts of
Vitaliy Feyer et al.
The journal of physical chemistry. A, 115(26), 7722-7733 (2011-05-20)
Core-level X-ray photoemission and near-edge X-ray absorption fine structure spectra of 5-methylcytosine, 5-fluorocytosine, and isocytosine are presented and discussed with the aid of high-level ab initio calculations. The effects of the methylation, halogenation, and isomerization on the relative stabilities of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service