Documents
HS81121B
Inoculating Needle/Loops
loop volume 1 μL, Rigid, sterile, blue
About This Item
UNSPSC Code:
41121800
NACRES:
NB.71
material
blue
sterility
sterile
feature
Rigid
packaging
pack of 1000 ea
manufacturer/tradename
Heathrow Scientific HS81121B
loop volume
1 μL
General description
The Inoculating Needle/Loops combination provides choice to spread or pick colonies and cultures. Transfer bacterial cultures without risk of contamination.
Features and Benefits
  • Use loop end to uniformly streak a gel surface
  • Use needle end for removal of colonies
  • Constructed of flexible polypropylene
  • Sterile, 20 per zip bag with lot-to-lot traceability
  • Disposable