Select a Size
€86.20
Estimated to ship on28 May 2025
Select a Size
About This Item
€86.20
Estimated to ship on28 May 2025
Recommended Products
material
polypropylene
purple
feature
to hold Side A 12 x 13 mm, Side B 18 x 13 mm, Side C 12 x 17 mm and side D 18 x 17 mm tubes
packaging
pack of 5 ea
manufacturer/tradename
Heathrow Scientific 120685
L × W × H
232 mm × 70 mm × 95 mm
Related Categories
General description
Features and Benefits
- Accommodates all brands of 13 and 16 mm round, flat and conical bottom blood collection & culture tubes
- Only 4-way rack available with a six way connection, link different or the same well sizes, end to end or side by side, in any orientation
- Customize the rack to fit the number of wells required, infinite units can be linked together or use one standalone unit
- Uniformly spaced wells help eliminate pipetting errors which enables more accurate results
- Molded alphanumeric identification for quick location and retrieval of tubes
- Robust, made from polypropylene that is chemical resistant and easy to clean
- Racks can be used in water baths and placed in freezers
- Assorted color pack available for color coding options
- Patent Pending
- Autoclavable
Choose from one of the most recent versions:
Certificates of Analysis (COA)