Skip to Content
Merck
All Photos(8)

Key Documents

HPA025736

Sigma-Aldrich

Anti-ACAT2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Acetyl-CoA acetyltransferase, Anti-Acetyl-CoA transferase-like protein, Anti-Cytosolic acetoacetyl-CoA thiolase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€541.00

€541.00


Available to ship on14 April 2025Details



Select a Size

Change View
100 μL
€541.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€541.00


Available to ship on14 April 2025Details


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LVSTRKGLIEVKTDEFPRHGSNIEAMSKLKPYFLTDGTGTVTPANASGINDGAAAVVLMKKSEADKRGLT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACAT2(39)

General description

The gene ACAT2 (acetyl-CoA acetyltransferase 2) is mapped to human chromosome 6q25.3. It is a transmembrane enzyme. It is mainly expressed in enterocytes in the small intestine and hepatocytes.

Immunogen

Acetyl-CoA acetyltransferase, cytosolic recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-ACAT2 antibody produced in rabbit has been used for western blotting.

Biochem/physiol Actions

ACAT2 (acetyl-CoA acetyltransferase 2) is responsible for the production of cholesteryl esters from cholesterol and fatty acids. In mouse model, absence of intestinal or hepatic ACAT2 results in atheroprotective effects. Thus, it is suggested as a target for treatment of hypercholesterolemia. Mutations in this gene are associated with changes in plasma lipid levels and coronary artery disease (CAD) risk.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77420

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ketone body utilization drives tumor growth and metastasis.
Martinez-Outschoorn UE
Cell Cycle, 11, 3964-3971 (2012)
Prioritization and association analysis of murine-derived candidate genes in anxiety-spectrum disorders.
Hettema JM
Biological Psychiatry, 70, 888-896 (2011)
ACAT2 and human hepatic cholesterol metabolism: identification of important gender-related differences in normolipidemic, non-obese Chinese patients.
Parini P, et. al.
Atherosclerosis, 207, 266-271 (2009)
Paolo Parini et al.
Atherosclerosis, 207(1), 266-271 (2009-05-27)
ACAT2 is a major cholesterol esterification enzyme specifically expressed in hepatocytes and may control the amount of hepatic free (unesterified) cholesterol available for secretion into bile or into HDL. This study aims to further elucidate physiologic roles of ACAT2 in
Xuelian He et al.
Gene, 566(2), 236-241 (2015-04-29)
Acyl-coenzyme A:cholesterol acyltransferase 2 (ACAT2) plays a critical role in the formation of cholesteryl esters from cholesterol and fatty acids, and is a potential target for treating hypercholesterolemia. We recently reported the significant effects of two human ACAT2 gene polymorphisms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service