Skip to Content
Merck
All Photos(5)

Key Documents

HPA023311

Sigma-Aldrich

Anti-SLC28A3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Concentrative Na(+, Anti-Solute carrier family 28 member 3, Anti-hCNT3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€541.00

€541.00


Available to ship on02 April 2025Details



Select a Size

Change View
100 μL
€541.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€541.00


Available to ship on02 April 2025Details


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

MELRSTAAPRAEGYSNVGFQNEENFLENENTSGNNSIRSRAVQSREHTNTKQDEEQVTVEQDSPRNREHMEDDDEEMQQKGCLERRYDTVCGFCRKHKTTLRH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC28A3(64078)

General description

The gene SLC28A3 (solute carrier family 28 member 3) is mapped to human chromosome 9q22. It is expressed in the pancreas, bone marrow, trachea, mammary gland, liver, prostate, intestine, brain and heart. The protein has 13 putative transmembrane helices.

Immunogen

Solute carrier family 28 member 3 recombinant protein epitope signature tag (PrEST)

Application

Anti-SLC28A3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting.[1] To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

SLC28A3 (solute carrier family 28 member 3) is a Na+-dependent purine and pyrimidine nucleoside symporter. It also transports nucleoside drugs and analogs.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75461

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrian Stecula et al.
Biochemistry, 56(27), 3475-3483 (2017-07-01)
Many anticancer and antiviral drugs are purine or pyrimidine analogues, which use membrane transporters to cross cellular membranes. Concentrative nucleoside transporters (CNTs) mediate the salvage of nucleosides and the transport of therapeutic nucleoside analogues across plasma membranes by coupling the
Takashi Yamamoto et al.
European journal of pharmacology, 557(1), 1-8 (2006-12-05)
We investigated the mechanism of the transport of ribavirin (1-beta-D-ribofuranosyl-1,2,4-trizole-3-carboxamide) into placental epithelial cells using human choriocarcinoma (BeWo) cells and Xenopus oocytes expressing human nucleoside transporters. In BeWo cells, when a relatively low concentration (123 nM) of ribavirin was used
M W Ritzel et al.
The Journal of biological chemistry, 276(4), 2914-2927 (2000-10-18)
The human concentrative (Na(+)-linked) plasma membrane transport proteins hCNT1 and hCNT2 are selective for pyrimidine nucleosides (system cit) and purine nucleosides (system cif), respectively. Both have homologs in other mammalian species and belong to a gene family (CNT) that also
Huankai Hu et al.
Molecular pharmacology, 69(5), 1542-1553 (2006-02-01)
We characterized the electrophysiology, kinetics, and quantitative structure-activity relationship (QSAR) of the human concentrative nucleoside transporter 3 (hCNT3) expressed in Xenopus laevis oocytes by measuring substrate-induced inward currents using a two-microelectrode voltage-clamp system. At membrane potentials between -30 and -150
Nicolas Skrypek et al.
Oncotarget, 6(13), 10853-10867 (2015-04-22)
Pancreatic adenocarcinoma (PDAC) is one of the most deadly cancers because of a lack of early diagnostic markers and efficient therapeutics. The fluorinated analog of deoxycytidine, gemcitabine and emerging FOLFIRINOX protocol (5-fluorouracil (5-FU), irinotecan/SN-38, oxaliplatin and leucovorin) are the main

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service