Skip to Content
Merck
All Photos(2)

Key Documents

HPA018119

Sigma-Aldrich

Anti-MARVELD2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DFNB49, Anti-FLJ30532, Anti-MARVEL domain containing 2, Anti-MRVLDC2, Anti-TRIC

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€541.00

€541.00


In StockDetails



Select a Size

Change View
100 μL
€541.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€541.00


In StockDetails


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:1000- 1:2500

immunogen sequence

AKYPVIQTDDERERYKAVFQDQFSEYKELSAEVQAVLRKFDELDAVMSRLPHHSESRQEHERISRIHEEFKKKKNDPTFLEKKERCDYLKNKLSHIKQRIQEYDKVMNWDV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

MARVEL domain containing 2 (MARVELD2) is a transmembrane protein located at the junction where three epithelial cells meet. It is known as tricellulin or tricellular tight junction protein. It possesses four transmembrane domains and the MAL and related proteins for vesicle trafficking and membrane link (MARVEL) domain. MARVELD2 is expressed in pancreatic ducts.[1]

Immunogen

MARVEL domain containing 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Mutations in the gene encoding MARVEL domain containing 2 (MARVELD2) are linked to autosomal recessive non-syndromic hearing loss (DFNB49).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73268

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cibelle Mariano et al.
European journal of cell biology, 90(10), 787-796 (2011-08-27)
Tight junctions are elaborate networks of transmembrane and cytosolic proteins that regulate epithelial permeability. Tricellulin was the first tight junction protein found at tricellular tight junctions, the specialized structures occurring where three cells meet together. Here, we summarize the current
Akira Takasawa et al.
Cell and tissue research, 351(1), 73-84 (2012-10-18)
Tricellulin is an important component of tricellular tight junctions (TJs) and is involved in the formation of tricellular contacts. However, little is known about its regulation during the assembly and disassembly of tricellular TJs. By using the well-differentiated pancreatic cancer
Aron Somorácz et al.
Pathology oncology research : POR, 20(4), 755-764 (2014-03-22)
Numerous data suggest that altered expression of tight junction proteins such as occludin and claudins plays important role in carcinogenesis. However, little is known about tricellulin, a transmembrane tight junction protein concentrated where three epithelial cells meet. We aimed to
Gowri Nayak et al.
Human genetics, 134(4), 423-437 (2015-02-11)
Pathogenic mutations of MARVELD2, encoding tricellulin, a tricelluar tight junction protein, cause autosomal recessive non-syndromic hearing loss (DFNB49) in families of Pakistan and Czech Roma origin. In fact, they are a significant cause of prelingual hearing loss in the Czech

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service