Skip to Content
Merck
All Photos(6)

Key Documents

HPA008704

Sigma-Aldrich

Anti-MRVI1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-JAW1-related protein isoform b antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€541.00

€541.00


Available to ship on15 April 2025Details



Select a Size

Change View
100 μL
€541.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€541.00


Available to ship on15 April 2025Details


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

NLVGLKLPDLSEAAEQEKGLPSELSPAIEEEESKSGLDVMPNISDVLLRKLRVHRSLPGSAPPLTEKEVENVFVQLSLAFRNDSYTLESRINQAERERNL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MRVI1(10335)

General description

MRVI1 (murine retrovirus integration site 1 homolog) gene is localized to human chromosome 11p15, which produces two alternatively spliced isoforms called MRVI1a and MRVI1b. They are both composed of an α-helical coiled coil domain, a hydrophobic C-terminal region, a basic N-terminal leucine zipper motif, multiple phosphorylation and N-linked glycosylation sites, and an ATP-binding region. It is localized to the endoplasmic reticulum (ER) membrane, and is a type-2 integral membrane protein.

Immunogen

JAW1-related protein isoform b recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MRVI1 (murine retrovirus integration site 1 homolog) acts as a substrate for the enzyme cGMP-dependent kinase-1 (PKG1). It is essential for motility, where it is responsible for the release of cyclic Ca2+ in a PKG1β-dependent manner. Any alterations in the inositol-1,4,5-trisphosphate receptor-1 (IP3R1)-MRVI1 system results in dysfunctional osteoclasts. In the smooth muscles of colon, this protein plays a key role in the NO/cGK-dependent inhibition of InsP3-dependent Ca2+-signaling.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71643

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ralph M Fritsch et al.
The Journal of biological chemistry, 279(13), 12551-12559 (2004-01-20)
Nitric oxide (NO)-mediated relaxation of colonic smooth muscle is crucial for the maintenance of human gut function. The molecular mechanisms of NO-dependent smooth muscle relaxation involve cyclic GMP-mediated inhibition of store-dependent calcium signaling. Recently, IRAG (inositol 1,4,5-trisphophate receptor-associated cGMP kinase
Sarabeth M Graham et al.
The AFCS-nature molecule pages, 2008, doi:10-doi:10 (2008-01-01)
Inositol 1, 4, 5-trisphosphate (IP3) receptor associated cyclic GMP (cGMP) kinase substrate (IRAG, also known as Mrv1) is a type-2 integral membrane endoplasmic reticulum (ER) protein, which interacts with IP3 Receptor type 1 (IP3R1), cGMP kinase I-β (cGKI β) and
Beatrice B Yaroslavskiy et al.
Laboratory investigation; a journal of technical methods and pathology, 90(10), 1533-1542 (2010-06-23)
Osteoclast activity is central to balanced bone turnover to maintain normal bone mass. A specialized osteoclast attachment to bone localizes acid secretion to remove bone mineral; in some cases, attachment is functionally impaired despite normal attachment proteins. The inositol-1,4,5-trisphosphate receptor-1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service