Skip to Content
Merck
All Photos(2)

Key Documents

HPA007420

Sigma-Aldrich

Anti-CDK3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cell division protein kinase 3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

PSEDTWPGVTQLPDYKGSFPKWTRKGLEEIVPNLEPEGRDLLMQLLQYDPSQRITAKTALAHPYFSSPEPSPAARQYVLQRFRH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDK3(1018)

Immunogen

Cell division protein kinase 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CDK3 (cyclin-dependent kinase 3) gene encodes a protein belonging to the cyclin-dependent protein kinase family. It is essential for the G1-S transition in mammalian cells. It is involved in the activation of E2F transcription factor 1, 2 and 3. This facilitates the entry of cells into S phase. It associates with cyclin C and phosphorylates pRb during the G0/G1 transition. This phosphorylation facilitates the cells to exit G0 state. It also phosphorylates activating transcription factor 1 (ATF1) and promotes cell proliferation and transformation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71411

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Hofmann et al.
Genes & development, 10(7), 851-861 (1996-04-01)
The cyclin-dependent kinases cdk2 and cdk3 are required for the G1-S transition in mammalian cells. Here we show that G1 arrest induced by the corresponding dominant-negative mutants of these enzymes, cdk2dn or cdk3dn, is resistant to the action of SV40
Duo Zheng et al.
Cancer research, 68(18), 7650-7660 (2008-09-17)
Cyclin-dependent kinase (cdk)-3, a member of the cdk family of kinases, plays a critical role in cell cycle regulation and is involved in G(0)-G(1) and G(1)-S cell cycle transitions. However, the role of cdk3 in cell proliferation, as well as
Shengjun Ren et al.
Cell, 117(2), 239-251 (2004-04-16)
G0 is a physiological state occupied by resting or terminally differentiated cells that have exited the cell cycle. In contrast to the well-characterized cyclin/cdk-mediated inactivation of pRb that controls the G1/S transition, little is known about regulation of the G0/G1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service