Skip to Content
Merck
All Photos(2)

Key Documents

HPA005478

Sigma-Aldrich

Anti-EZH1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ENX-2, Anti-Enhancer of zeste homolog 1, Anti-Histone-lysine N-methyltransferase EZH1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect immunofluorescence: suitable

immunogen sequence

MGDEVKEEDETFIEELINNYDGKVHGEEEMIPGSVLISDAVFLELVDALNQYSDEEEEGHNDTSDGKQDDSKEDLPVTRKRKRHAIEGNKKSSKKQFPNDMIFSAIASMFPENGVPDDMKERYRELTEMSDPNALPPQCTPNIDGPNA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EZH1(2145)

General description

Enhancer of zeste homolog 1 (EZH1) is a part of non-canonical PRC2 (Polycomb repressive complex), and is an H3K27 methyltransferase. It localizes to chromatin. It is expressed ubiquitously, with high expression profiles in skeletal muscle, brain and kidney. It is expressed in adult and fetal tissues as two major transcripts. EZH1 protein is composed of 747 amino acids. In humans, this gene is located on chromosome 17.

Immunogen

enhancer of zeste 1 polycomb repressive complex 2 subunit

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Enhancer of zeste homolog 1 (EZH1) mediates PRC2 complex activity, which in turn is responsible for the induction and maintainance of gene respression. PRC2 is recruited to chromatin, where EZH1 helps it to catalyze the trimethylation of histone H3 (H3K27me3), on its lysine 27 residue. This protein also promotes mRNA transcription by facilitating RNA polymerase II elongation. Since EZH1 is a histone methyltransferase, it maintains the undifferentiated state of hematopoietic stem cells (HSC) and prevents their senescence. Inactivation of this gene leads to degeneration of hair follicles as the proliferation of hair follicle stem cells is inhibited. In epidermal stem cells, EZH1 represses the differentiation of Merkel stem cell lineage.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86929

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K J Abel et al.
Genomics, 37(2), 161-171 (1996-10-15)
Recent transcription mapping efforts within chromosome 17q21 have led to the identification of a human homolog of the Drosophila gene Enhancer of zeste, E(z). A member of the Polycomb group (Pc-G) of proteins, Drosophila E(z) acts as a negative regulator
Kambiz Mousavi et al.
Molecular cell, 45(2), 255-262 (2011-12-27)
Polycomb group (PcG) proteins initiate the formation of repressed chromatin domains and regulate developmental gene expression. A mammalian PcG protein, enhancer of zeste homolog 2 (Ezh2), triggers transcriptional repression by catalyzing the addition of methyl groups onto lysine 27 of
Raphael Margueron et al.
Molecular cell, 32(4), 503-518 (2008-11-26)
Polycomb group proteins are critical to maintaining gene repression established during Drosophila development. Part of this group forms the PRC2 complex containing Ez that catalyzes di- and trimethylation of histone H3 lysine 27 (H3K37me2/3), marks repressive to transcription. We report
G Laible et al.
The EMBO journal, 16(11), 3219-3232 (1997-06-02)
Gene silencing is required to stably maintain distinct patterns of gene expression during eukaryotic development and has been correlated with the induction of chromatin domains that restrict gene activity. We describe the isolation of human (EZH2) and mouse (Ezh1) homologues
Evan S Bardot et al.
The EMBO journal, 32(14), 1990-2000 (2013-05-16)
While the Polycomb complex is known to regulate cell identity in ES cells, its role in controlling tissue-specific stem cells is not well understood. Here we show that removal of Ezh1 and Ezh2, key Polycomb subunits, from mouse skin results

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service