Skip to Content
Merck
All Photos(1)

Key Documents

HPA003186

Sigma-Aldrich

Anti-DDX53 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cancer-associated gene protein antibody produced in rabbit, Anti-DEAD box protein 53 antibody produced in rabbit, Anti-DEAD box protein CAGE antibody produced in rabbit, Anti-Probable ATP-dependent RNA helicase DDX53 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

immunogen sequence

DDLSSDFNIQGISAESLHGNSEQSDQERAVEDFKSGNIKILITTDIVSRGLDLNDVTHVYNYDFPRNIDVYVHRVGYIGRTGKTGTSVTLITQRDSKMAGELIKILDRANQSVPEDLVVMAEQYKLNQQKRHRETRSRKPGQRRKEFY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DDX53(168400)

Immunogen

Probable ATP-dependent RNA helicase DDX53 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

DDX53 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 53) is an intronless gene that encodes a protein containing domains similar to that of DEAD-box helicase proteins. The protein is 630 amino acids long and may function as a helicase. The gene is involved in tumorigenesis. The gene is also called as cancer-associated gene (CAGE) and is implicated in several types of cancer cells and may serve as a potential target for antitumor immunotherapy. It may also function in cellular proliferation. CAGE negatively regulates p53 expression and makes the cancerous cells resistant to anti-cancer drugs.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bomsoo Cho et al.
Biochemical and biophysical research communications, 292(3), 715-726 (2002-04-02)
We applied serological analysis of cDNA expression library technique to identify cancer-associated genes. We screened cDNA expression libraries of human testis and gastric cancer cell lines with sera of patients with gastric cancers. We identified a gene whose expression is
Youngmi Kim et al.
The Journal of biological chemistry, 285(34), 25957-25968 (2010-06-11)
The role of the cancer/testis antigen CAGE in drug resistance was investigated. The drug-resistant human melanoma Malme3M (Malme3M(R)) and the human hepatic cancer cell line SNU387 (SNU387(R)) showed in vivo drug resistance and CAGE induction. Induction of CAGE resulted from
Youngmi Kim et al.
Journal of microbiology and biotechnology, 18(3), 600-610 (2008-04-05)
A cancer-associated antigen gene (CAGE) was identified by serological analysis of a recombinant cDNA expression library (SEREX). The gene was identified by screening cDNA expression libraries of human testis and gastric cancer cell lines with sera from patients with gastric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service