Skip to Content
Merck
All Photos(3)

Documents

HPA003175

Sigma-Aldrich

Anti-BACH1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BACH-1, Anti-BTBD24

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

immunogen sequence

EPLIQFAYTAKLILSKENVDEVCKCVEFLSVHNIEESCFQFLKFKFLDSTADQQECPRKKCFSSHCQKTDLKLSLLDQRDLETDEVEEFLENKNVQTPQCKLRRYQGNAKASPPLQDSASQTYESMCLEKDAALA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BACH1(571)

Immunogen

Transcription regulator protein BACH1 recombinant protein epitope signature tag (PrEST)

Application

Anti-BACH1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

BTB and CNC homology 1, basic leucine zipper transcription factor 1 is a protein encoded by the gene BACH1 in humans. It belongs to the DEAH helicase family. It is a basic leucine zipper protein containing broad complex, tramtrack, bric-a-brac/poxvirus and zinc finger (BTB/POZ) domains. It forms a heterodimer with MafK and functions as a hypoxia-inducible repressor for the HO-1 (Heme Oxygenase-1) gene and regulates oxygen homeostasis. This protein binds to the BRCT repeats of BRCA1 and facilitates its DNA repair mechanism.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84783.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiyoung Lee et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(3), E364-E373 (2014-01-08)
The sources and consequences of nongenetic variability in metastatic progression are largely unknown. To address these questions, we characterized a transcriptional regulatory network for the metastasis suppressor Raf kinase inhibitory protein (RKIP). We previously showed that the transcription factor BACH1
Tomomi Kitamuro et al.
The Journal of biological chemistry, 278(11), 9125-9133 (2003-01-04)
Heme oxygenase 1 (HO-1) catalyzes heme breakdown, eventually releasing iron, carbon monoxide, and bilirubin IXalpha. HO-1 is induced by its substrate heme and various environmental factors, which represents a protective response against oxidative stresses. Here we show that hypoxia represses
S B Cantor et al.
Cell, 105(1), 149-160 (2001-04-13)
BRCA1 interacts in vivo with a novel protein, BACH1, a member of the DEAH helicase family. BACH1 binds directly to the BRCT repeats of BRCA1. A BACH1 derivative, bearing a mutation in a residue that was essential for catalytic function
Mathias Uhlén et al.
Molecular & cellular proteomics : MCP, 11(3), M111-M111 (2011-11-02)
The Human Proteome Project has been proposed to create a knowledge-based resource based on a systematical mapping of all human proteins, chromosome by chromosome, in a gene-centric manner. With this background, we here describe the systematic analysis of chromosome 21

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service