Skip to Content
Merck
All Photos(1)

Key Documents

H4542

Sigma-Aldrich

Cytochrome P450 2C9*3 (I359L), human

recombinant, expressed in E. coli

Synonym(s):

Cytochrome P450, purified, soluble, CYP2C9 I359L, CYP2C9 Il359Leu

Sign Into View Organizational & Contract Pricing

Select a Size

1000 PMOL
€614.00

€614.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
1000 PMOL
€614.00

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54

€614.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in E. coli

description

purified, soluble

form

liquid

specific activity

3-56 units/mg protein

mol wt

54423.1 g/mol

concentration

3.5-25 mg protein/mL (protein)
50-150 nmol/mL
6-14 nmol/mg protein (specific content)

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CYP2C9(1559)

Looking for similar products? Visit Product Comparison Guide

General description

Supplied as purified soluble proteins, rather than microsomal preparations.

Unit Definition

One unit will convert 1.0 nanomole of diclofenac (2-(2,6-dichloranilino) phenylacetic acid) 4`-hydroxylated product per minute at pH 7.4 at 37 °C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service