Recommended Products
Assay
≥98% (TLC)
storage temp.
−20°C
SMILES string
NC(CCS(O)=O)C(O)=O
InChI
1S/C4H9NO4S/c5-3(4(6)7)1-2-10(8)9/h3H,1-2,5H2,(H,6,7)(H,8,9)
InChI key
PDNJLMZEGXHSCU-UHFFFAOYSA-N
Biochem/physiol Actions
Neurotransmitter candidate
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Neuron, 3(1), 129-132 (1989-07-01)
Three major subtypes of glutamate receptors that are coupled to cation channels are known. Recently an additional subtype that is coupled to G proteins and stimulates inositol phospholipid metabolism (the metabotropic glutamate receptor) has been proposed. The pharmacological characteristics of
Homocysteic acid, an endogenous agonist of NMDA-receptor: release, neuroactivity and localization.
Advances in experimental medicine and biology, 203, 253-262 (1986-01-01)
The Journal of physiology, 444, 441-457 (1991-12-01)
1. The effect of sulphur-containing analogues of glutamate and aspartate on the membrane current of glial cells was studied by whole-cell clamping Müller cells isolated from the salamander retina. 2. L-Cysteic acid (CA), L-cysteinesulphinic acid (CSA), L-homocysteic acid (HCA), L-homocysteinesulphinic
L-homocysteine sulfinic acid and L-homocysteic acid stimulate phosphoinositide hydrolysis in rat cortical neurons.
Annals of the New York Academy of Sciences, 1003, 461-463 (2003-12-20)
Biology of reproduction, 74(3), 487-491 (2005-11-18)
In previous work, variation in sperm mobility phenotype was attributed to the proportion of ejaculated fowl sperm containing dysfunctional mitochondria. In the present work, latent mitochondrial dysfunction was inferred from patterns of sperm egress from the oviduct's sperm-storage tubules. In
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service