Skip to Content
Merck
All Photos(1)

Key Documents

H3288

Sigma-Aldrich

HEMADO

≥99% (HPLC), solid

Synonym(s):

2-(1-Hexynyl)-N6-methyladenosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23N5O4
CAS Number:
Molecular Weight:
361.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99% (HPLC)

form

solid

color

white

solubility

DMSO: 16 mg/mL

storage temp.

2-8°C

SMILES string

CCCCC#Cc1nc(NC)c2ncn([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)c2n1

InChI

1S/C17H23N5O4/c1-3-4-5-6-7-11-20-15(18-2)12-16(21-11)22(9-19-12)17-14(25)13(24)10(8-23)26-17/h9-10,13-14,17,23-25H,3-5,8H2,1-2H3,(H,18,20,21)/t10-,13-,14-,17-/m1/s1

InChI key

KOCIMZNSNPOGOP-IWCJZZDYSA-N

Biochem/physiol Actions

A3 adenosine receptor agonist

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosaria Volpini et al.
Journal of medicinal chemistry, 45(15), 3271-3279 (2002-07-12)
A series of N(6)-alkyl-2-alkynyl derivatives of adenosine (Ado) have been synthesized and evaluated for their affinity at human A(1), A(2A), and A(3) receptors and for their potency at A(2B) adenosine receptor subtypes. The corresponding 2-(1-alkynyl) derivatives of 5'-N-ethylcarboxamidoadenosine (NECA) and
Kerry Barkan et al.
Scientific reports, 10(1), 20781-20781 (2020-11-29)
The adenosine A3 receptor (A3R) belongs to a family of four adenosine receptor (AR) subtypes which all play distinct roles throughout the body. A3R antagonists have been described as potential treatments for numerous diseases including asthma. Given the similarity between
Leigh A Stoddart et al.
British journal of pharmacology, 171(16), 3827-3844 (2014-04-23)
The highly conserved tryptophan (W6.48) in transmembrane domain 6 of GPCRs has been shown to play a central role in forming an active conformation in response to agonist binding. We set out to characterize the effect of this mutation on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service