Skip to Content
Merck
All Photos(1)

Key Documents

H3042

Sigma-Aldrich

Hemopressin trifluoroacetate salt

≥98% (HPLC), powder

Synonym(s):

Hemopressin from rat, Hemopressin from rat brain, L-Prolyl-L-valyl-L-asparaginyl-L-phenylalanyl-L-lysyl-L-phenylalanyl-L-leucyl-L-seryl-L-histidine trifluoroacetate salt, Pro-Val-Asn-Phe-Lys-Phe-Leu-Ser-His trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€95.20

€95.20


Estimated to ship on07 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€95.20

About This Item

Empirical Formula (Hill Notation):
C53H77N13O12 · xC2HF3O2
CAS Number:
Molecular Weight:
1088.26 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€95.20


Estimated to ship on07 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

H2O: ≥2 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC(C)C[C@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@H](CCCCN)NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](CC(N)=O)NC(=O)[C@@H](NC(=O)[C@@H]3CCCN3)C(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc4c[nH]cn4)C(O)=O

InChI

1S/C53H77N13O12.C2HF3O2/c1-30(2)22-37(47(71)65-42(28-67)51(75)64-41(53(77)78)25-34-27-56-29-58-34)60-49(73)39(24-33-16-9-6-10-17-33)61-46(70)36(18-11-12-20-54)59-48(72)38(23-32-14-7-5-8-15-32)62-50(74)40(26-43(55)68)63-52(76)44(31(3)4)66-45(69)35-19-13-21-57-35;3-2(4,5)1(6)7/h5-10,14-17,27,29-31,35-42,44,57,67H,11-13,18-26,28,54H2,1-4H3,(H2,55,68)(H,56,58)(H,59,72)(H,60,73)(H,61,70)(H,62,74)(H,63,76)(H,64,75)(H,65,71)(H,66,69)(H,77,78);(H,6,7)/t35-,36-,37-,38-,39-,40-,41-,42-,44-;/m0./s1

InChI key

FXBODCREWDDYNR-DFZCIRLNSA-N

Biochem/physiol Actions

Hemopressin is a novel endogenous hypotensive peptide and a CB1 inverse agonist.
Hemopressin is an endogenous hypotensive nonapeptide derived from hemoglobin. Hemopressin reduces appetite, acting as a cannabinoid CB1 inverse agonist. It blocks signaling by CB(1) receptors but not by other members of family A G protein-coupled receptors (including the closely related CB(2) receptors.

Features and Benefits

This compound is featured on the Cannabinoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to cannabinoid receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service