biological source
human
Quality Level
recombinant
expressed in E. coli
form
liquid
concentration
1 mg/mL
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... H3.2(440686)
Looking for similar products? Visit Product Comparison Guide
Physical form
Supplied as 1 mg/mL in 20 mM NaPO4, pH 7.0, 0.3 M NaCl, 1 mM EDTA, and 1 mM DTT.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service