Skip to Content
Merck
All Photos(1)

Key Documents

H1763

Sigma-Aldrich

Hippuryl-Gly-Gly

Synonym(s):

N-Benzoyl-Gly-Gly-Gly

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15N3O5
CAS Number:
Molecular Weight:
293.28
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥99% (HPLC)

form

powder

solubility

1 M NH4OH: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

OC(=O)CNC(=O)CNC(=O)CNC(=O)c1ccccc1

InChI

1S/C13H15N3O5/c17-10(15-8-12(19)20)6-14-11(18)7-16-13(21)9-4-2-1-3-5-9/h1-5H,6-8H2,(H,14,18)(H,15,17)(H,16,21)(H,19,20)

InChI key

CYQZRSVDAIOAIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Substrates

Substrate for angiotensin converting enzyme (ACE)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Mor et al.
Journal of pharmacological methods, 23(2), 141-153 (1990-04-01)
We have developed a pressure-dependent isolated lung perfusion system that can be used for the determination of pulmonary enzyme activity and kinetics under physiologic conditions. This development was done using two different artificial radiolabeled substrates, glycine-1-hippuryl-L-histidyl-L-leucine and phenyl-4(n)-hippuryl-glycyl-glycine, for the
Simple radioassay for measuring serum activity of angiotensin-converting enzyme in sarcoidosis.
P K Rohatgi et al.
Chest, 78(1), 69-76 (1980-07-01)
T A Stewart et al.
Peptides, 2(2), 145-152 (1981-01-01)
We purified peptidyl-dipeptidase (converting enzyme, EC 3.4.15.1) to homogeneity from the membrane fraction of human lung and for comparison, from human and hog kidney. The membrane-bound lung enzyme was purified 1800-fold with 19% yield, and the kidney enzyme 640-fold with
Anick Vandingenen et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 132(1), 27-35 (2002-05-09)
Angiotensin converting enzyme (ACE) activity, defined as a captopril-inhibitable dipeptidyl carboxypeptidase activity towards 3H-hippurylglycylglycine, was demonstrated in haemolymph, testes and ovaries of the grey fleshfly Neobellieria bullata, hereby suggesting a physiological role for ACE in these particular tissues. While the
C E Odya et al.
Canadian journal of physiology and pharmacology, 64(1), 50-54 (1986-01-01)
Bradykinin and 22 of its analogs were evaluated for their abilities to inhibit the hydrolysis of [3H]hippurylglycylglycine by purified porcine kidney angiotensin I converting enzyme. The mean inhibitory concentration (IC50) for bradykinin was 1.2 +/- 0.2 X 10(-6) M. Except

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service