Skip to Content
Merck
All Photos(1)

Key Documents

GN724201

Greiner universal beaker

volume 100 mL, size 56 mm × 70 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121803
Pricing and availability is not currently available.

packaging

pack of 1200 ea

manufacturer/tradename

Greiner 724201

availability

available only in China, China

size

56 mm × 70 mm

volume

100 mL

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all glass beakers

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service