Skip to Content
Merck
All Photos(1)

Documents

GN680065

Greiner CELLMASTER roller bottle

850 cm2, 122 x 271 mm, 2.5 L, TC treated, sterile

Synonym(s):

Greiner CELLMASTER roller bottles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122105

material

clear bottle
narrow-mouth bottle
polystyrene bottle
round bottle

sterility

sterile

feature

autoclavable: no
cap
graduations
ID field
short smooth surface, TC-treated

packaging

pack of 24 ea

manufacturer/tradename

Greiner 680065

availability

available only in China

capacity

2,550 mL

diam. × H

122 mm × 271 mm

Looking for similar products? Visit Product Comparison Guide

General description

Greiner CELLMASTER roller bottles are sterile and tissue culture treated, thus supporting the growth of attached cells, also known as anchorage-dependent cell lines. In addition, these bottles are non-pyrogenic and certified DNase/RNase free as well as being very sturdy and resistant to cracking.


Material:
Bottle: Polyethylene Terephthalate
Cap: High Densiity Polyethylene

Legal Information

CELLMASTER is a trademark of Greiner Bio-One GmbH

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service