Skip to Content
Merck
All Photos(1)

Documents

GERPN3244

ECL Start Western Blotting Detection Reagent

Cytiva RPN3244, pkg of 400 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105332
NACRES:
NA.32

packaging

pkg of 400 mL

manufacturer/tradename

Cytiva RPN3244

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Amersham ECL start is a chemiluminescent detection reagent in the Amersham ECL product family that is recommended for confirmatory analysis and detection of high to medium levels of endogenous proteins. Amersham ECL start has a long signal duration, which enables multiple exposures of the membrane, plus a convenient time window between experiment and analysis.

Features and Benefits

  • Flexibility: Long signal duration provides assay design flexibility and robust detection of large experimental sets.
  • Low background: Produces low background, regardless of the method of detection.
  • Stability of the working solution: The working solution is stable for 5 days.
  • Long shelf life: A long shelf life of 18 months is convenient for those who run Western blots infrequently.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

ECL is a trademark of Cytiva

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service