Skip to Content
Merck
All Photos(2)

Key Documents

GERPN203D

Hybond® ECL nitrocellulose membranes

W × L 20 cm × 3 m, roll nitrocellulose, pack of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

material

nitrocellulose
roll nitrocellulose

form

solid

feature

wetted part: no

packaging

pack of 1 ea

manufacturer/tradename

Cytiva RPN203D

W × L

20 cm × 3 m

color

White

pore size

0.45 μm

Looking for similar products? Visit Product Comparison Guide

Legal Information

ECL is a trademark of Cytiva
Hybond is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service