GE80-6444-23
Blot processing tray pack for Processor Plus
pack of 1 kit
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.22
feature
wetted part: no
packaging
pack of 1 kit
manufacturer/tradename
Cytiva 80644423
parameter
4-40 °C temp. range
General description
Blot Processing Tray Pack
Processor Plus is a novel fluid delivery system that automates acrylamide gel staining and western blot processing. Changing from one application to the other is achieved by a fast, simple switch of a tray.
Processor Plus is a novel fluid delivery system that automates acrylamide gel staining and western blot processing. Changing from one application to the other is achieved by a fast, simple switch of a tray.
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)