Skip to Content
Merck
All Photos(1)

Documents

GE28-9440-06

Protein A Mag Sepharose

Cytiva 28-9440-06, pack of 1 × 500 μL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

ligand

native protein A

packaging

pack of 1 × 500 μL

manufacturer/tradename

Cytiva 28-9440-06

storage condition

(20% Ethanol, 20% medium slurry)

parameter

Room temperature temp. range

matrix

paramagnetic, spherical, highly cross-linked agarose particles

capacity

8- 17 mg binding capacity(human IgG/ml gel)

storage temp.

2-8°C

Related Categories

General description

Protein A Mag Sepharose are magnetic beads designed to simplify enrichment of target proteins by immunoprecipitation or pull-down applications. Based on Sepharose with native Protein A as ligand.

Application

Protein A Mag Sepharose are magnetic beads designed to simplify enrichment of target Proteins by immunoprecipitation or pull-down applications. The magnetic beads are based on Sepharose with native Protein A as ligand. Protein A has high affinity for the Fc region of IgG antibodies from a broad variety of species.

There are two protocols available for Protein enrichment: the cross-link protocol and the classic protocol. The protocols are simple with elution conditions optimized for both electrophoresis and MS analysis.

Protein A Mag Sepharose combines well-proven enrichment methods with the magnetic beads platform, which is highly suitable for small-scale sample preparation. Together with MagRack 6, a separation tool for six microcentrifuge tubes, up to six samples can be processed in parallel.

Features and Benefits

  • Visible and dense Sepharose based magnetic beads with Protein A ligand, easy to spot and collect bound target Protein
  • Non-adherent beads eliminate smearing effects and aggregate formation. Can be used without detergents
  • Simple capture of target Protein in small or large sample volumes (Low microliter to high milliliter scale)
  • Optimized capacity for enrichment/immunoprecipitation of target Proteins - only Low amounts of antibody needed
  • Flexible protocols with elution conditions optimized for both electrophoresis and MS analysis

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows various purification options for Protein A Sepharose chromatography media and describes typical binding and elution conditions for Protein A Sepharose chromatography media.

This page describes immunoprecipitation (immunoaffinity or pull-down techniques).

Protocols

Protein A Mag Sepharose® Xtra and Protein G Mag Sepharose® Xtra enable high-capacity small-scale antibody purification and screening.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service