Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9425-44

Tripleprep

Cytiva 28-9425-44, sufficient for 50 purifications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.55
Pricing and availability is not currently available.

usage

sufficient for 50 purifications

manufacturer/tradename

Cytiva 28-9425-44

Related Categories

General description

The illustra triplePrep Kit is designed for rapid, simultaneous extraction and isolation of high quality genomic DNA (gDNA), total RNA, and total denatured proteins from single sample of animal tissue and mammalian cells.

Application

The isolated gDNA, total RNA, and total denatured proteins are suitable for genomic and proteomic applications such as PCR, restriction enzyme digestion, sequencing, array CGH, RT-PCR, gene expression microarray, SDS-PAGE, Western blotting, 2-D DIGE, and LCMS.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

illustra is a trademark of Cytiva

Pictograms

Environment

Signal Word

Danger

Hazard Statements

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows downstream applications after genomic DNA purification.

Protocols

This page shows genomic DNA purification from sample on FTA elute from Cytiva.

Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service