Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0686-01

Sigma-Aldrich

Mini Q PC 3.2/3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

ligand

-OCH2CH(OH)CH2OCH2CH(OH)CH2N+(CH3)3

description

Ion Exchanger Type (value)

packaging

pkg of 1 ea

parameter

100 bar (1450 psi) (Over the Packed Bed During Operation)
1450 psi

bed size

3.2 mm × 30 mm

bed volume

0.24 mL

column I.D.

3.2 mm

matrix

PS-DVB particle platform
monodisperse polystyrene/divinyl benzene particles

matrix active group

ammonium ion, quaternary phase

particle size

3 μm

cleaning

2-14

working range

3-11

separation technique

anion exchange

General description

Mini Q PC 3.2/3 Precision Column (0.24 ml) is prepacked with MiniBeads for microscale final polishing and analytical ion exchange separations.

Application

Precision Column Holder enables use of precision columns with high-performance chromatography systems such as ÄKTA® design.

Features and Benefits

  • Monodisperse 3 μm nonporous beads yield exceptional resolution, speed, reproducibility, and durability.
  • PC columns are designed for micropurification.
  • Nonporous matrix of MiniBeads improves resolution and flow kinetics, better withstands back pressure, and minimizes nonspecific adsorption of the sample.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Mini Q is a trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service