Skip to Content
Merck
All Photos(1)

Key Documents

G7378

Sigma-Aldrich

Glycinamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CONH2 · HCl
CAS Number:
Molecular Weight:
110.54
Beilstein:
3554199
EC Number:
MDL number:
UNSPSC Code:
12352200

pH

7.4-8.8

pKa (25 °C)

8.1

pKa (20 °C)

8.20

SMILES string

Cl.NCC(N)=O

InChI

1S/C2H6N2O.ClH/c3-1-2(4)5;/h1,3H2,(H2,4,5);1H

InChI key

WKNMKGVLOWGGOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gottfried K Schroeder et al.
Biochemistry, 46(13), 4037-4044 (2007-03-14)
As a model for mechanistic comparison with peptidyl transfer within the ribosome, the reaction of aqueous glycinamide with N-formylphenylalanine trifluoroethyl ester (fPhe-TFE) represents an improvement over earlier model reactions involving Tris. The acidity of trifluoroethanol (pKa 12.4) resembles that of
Xiaodong Jia et al.
The Journal of organic chemistry, 78(18), 9450-9456 (2013-08-21)
A catalytic α-sp(3) C-H oxidation of peptides and glycine amides was achieved under radical cation salt catalysis in the presence of O2, producing a series of substituted quinolines. The scope of this reaction shows good functional group tolerance and high
Eric Loeser et al.
Analytical chemistry, 79(14), 5382-5391 (2007-05-29)
When mobile-phase salt content is increased, cationic analytes often show increased retention. This effect is generally attributed to chaotropic or ion pairing effects. However, a cation exclusion mechanism could explain the same effects. In this study, experimental conditions were manipulated
Robert J Cherney et al.
Bioorganic & medicinal chemistry letters, 20(8), 2425-2430 (2010-03-30)
We describe the design, synthesis, and evaluation, of gamma-lactams as glycinamide replacements within a series of di- and trisubstituted cyclohexane CCR2 antagonists. The lactam-containing trisubstituted cyclohexanes proved to be more potent than the disubstituted analogs, as trisubstituted analog, lactam 13
Brett C Bookser et al.
Journal of medicinal chemistry, 48(24), 7808-7820 (2005-11-24)
4-(Phenylamino)-5-phenyl-7-(5-deoxy-beta-D-ribofuranosyl)pyrrolo[2,3-d]pyrimidine 1 and related compounds known as "diaryltubercidin" analogues are potent inhibitors of adenosine kinase (AK) and are orally active in animal models of pain such as the rat formalin paw model (GP3269 ED50= 6.4 mg/kg). However, the utility of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service