Skip to Content
Merck
All Photos(1)

Key Documents

G5510

Sigma-Aldrich

Glycogen azure

≥98% (HPLC), from rabbit liver, suitable for substrate for α-amylase

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€358.00

€358.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€358.00

About This Item

MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79

€358.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

rabbit liver

Quality Level

Assay

≥98% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

blue

suitability

suitable for substrate for α-amylase

storage temp.

2-8°C

Application

Dye-labeled carrier that can be used for visualizing alcohol-precipitated nucleic acid pellets.
Glycogen azure was used in DNA precipitation. [1]

Other Notes

Rabbit liver glycogen covalently linked with Remazol Brilliant Blue R.
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Preparation Note

Prepared by a modification of the procedure of Rinderknecht, H., et al., Experientia, 23, 805 (1967).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mary Waltner-Law et al.
The Journal of biological chemistry, 278(12), 10427-10435 (2003-01-18)
Although many genes are regulated by the concerted action of several hormones, hormonal signaling to gene promoters has generally been studied one hormone at a time. The phosphoenolpyruvate carboxykinase (PEPCK) gene is a case in point. Transcription of this gene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service