Skip to Content
Merck
All Photos(1)

Key Documents

F9751

Sigma-Aldrich

N-Formyl-L-tyrosine

powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO4
CAS Number:
Molecular Weight:
209.20
UNSPSC Code:
12352200
PubChem Substance ID:

form

powder

color

white

storage temp.

−20°C

SMILES string

[H]C(=O)N[C@@]([H])(Cc1ccc(O)cc1)C(O)=O

InChI

1S/C10H11NO4/c12-6-11-9(10(14)15)5-7-1-3-8(13)4-2-7/h1-4,6,9,13H,5H2,(H,11,12)(H,14,15)/t9-/m0/s1

InChI key

ROUWPHMRHBMAFE-VIFPVBQESA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leo D Bemena et al.
Journal of biochemistry, 163(2), 123-131 (2018-01-25)
The outermost layer of the yeast Saccharomyces cerevisiae spore, termed the dityrosine layer, is primarily composed of bisformyl dityrosine. Bisformyl dityrosine is produced in the spore cytosol by crosslinking of two formyl tyrosine molecules, after which it is transported to
Mostafa Basiony et al.
Journal of fungi (Basel, Switzerland), 6(2) (2020-06-07)
The dityrosine layer is a unique structure present in the spore wall of the budding yeast Saccharomyces cerevisiae. The primary constituent of this layer is bisformyl dityrosine. A sporulation-specific protein, Dit1 is localized in the spore cytosol and produces a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service