F2385
N-Formyl-Met-Leu-Phe-Lys acetate salt
≥90% (HPLC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C27H43N5O6S
CAS Number:
Molecular Weight:
565.73
MDL number:
UNSPSC Code:
12352200
Assay
≥90% (HPLC)
storage temp.
−20°C
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Further studies on the structural requirements for synthetic peptide chemoattractants.
R J Freer et al.
Biochemistry, 19(11), 2404-2410 (1980-05-27)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service