EP6731010355
Eppendorf® CellXpert® C170i CO2 Incubator
![New Product new](/static/new_product_badge.png)
water/humidity monitoring, inner door with 4 door segments, handle left side, AC/DC input 120 V, US 2-pin plug (US/JP/South America)
All Photos(1)
About This Item
UNSPSC Code:
41104405
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 6731010355
General description
CellXpert® C170i, inner door with 4 door segments, handle left side, water and humidity monitoring package, 100 – 127 V/50 – 60 Hz (US/JP/South America)
Features and Benefits
- water and humidity monitoring package
- handle left side
- inner door with 4 door segments
- 180° HTD (standard with C170i)
- including water tray and four shelves (stainless steel)
- 100 – 127 V, 50 – 60 Hz
Legal Information
CellXpert is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG