EP5895177000
Eppendorf® Rotor Seal and Sealing Grease
for rotor FA-6x250
Synonym(s):
Eppendorf® Rotor Lid Seals
About This Item
Recommended Products
manufacturer/tradename
Eppendorf® 5895177000
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service