Skip to Content
Merck
All Photos(1)

Key Documents

EP5492008810

Eppendorf® Buckets

fitting rotors A-12-11 & A-8-11, set of 8 pieces

Synonym(s):

Eppendorf® Buckets

Sign Into View Organizational & Contract Pricing

Select a Size

8 EA
€54.40

€54.40


Check Cart for Availability


Select a Size

Change View
8 EA
€54.40

About This Item

UNSPSC Code:
41103910
NACRES:
NB.82

€54.40


Check Cart for Availability

feature

fitting rotors A-12-11 & A-8-11

packaging

set of 8 pieces

manufacturer/tradename

5492008810
Eppendorf® 5492008810
Eppendorf®

technique(s)

centrifugation: suitable

Looking for similar products? Visit Product Comparison Guide

General description

Buckets for Eppendorf centrifuge buckets.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service