Key Documents
EP0030625102
Eppendorf® Microplate 384/F-PP, 384 wells
white polypropylene, flat bottom clear wells, PCR clean, pkg of 240 ea
Select a Size
240 EA
€1,460.00
€1,460.00
Check Cart for Availability
Select a Size
Change View
240 EA
€1,460.00
About This Item
UNSPSC Code:
41122107
NACRES:
NB.15
material:
flat bottom clear wells
white polypropylene
size:
384 wells
€1,460.00
Check Cart for Availability
material
flat bottom clear wells
white polypropylene
packaging
pkg of 240 ea
manufacturer/tradename
Eppendorf® 0030625102
technique(s)
PCR: suitable
size
384 wells
General description
Experience a new level of transparency with Eppendorf Microplates. With the highest quality polypropylene and superior well design, you can pipette, control and recover samples with minimal effort.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)