Merck
All Photos(4)
Key Documents
EP0030129849
Eppendorf® BioBased twin.tec 96 Well PCR Plate
greener alternative
skirt, clear (with OptiTrack high contrast labelling)
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material:
clear (with OptiTrack high contrast labelling)
polypropylene
size:
96 well
sterility:
non-sterile
feature:
lid: no
skirt
Pricing and availability is not currently available.
material
clear (with OptiTrack high contrast labelling)
polypropylene
sterility
non-sterile
feature
lid: no
skirt
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 0030129849
greener alternative product characteristics
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
size
96 well
well volume
150 μL
suitability
suitable for PCR
greener alternative category
General description
Eppendorf® twin.tec PCR Plate 96, BioBased, skirted, PCR clean, clear, 25 plates
Features and Benefits
  • Reducing your environmental footprint inthe lab
  • Produced from second-generation biobased with 90% of thefeedstock attributed to renewableresources (i.e. waste and residues fromvegetable oil refining or waste cooking oil), avoiding the use of fossilfuel-based petroleum
  • Eppendorf production facility is ISCC PLUS (InternationalSustainability & Carbon Certification) certified, confirming the rawmaterials used at the facility are “bio-circular”. Enables traceability alongthe entire supply chain
  • Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
  • Unique laser-engraved optical guiding grid and OptiTrack matrix for quick orientation when pipetting manually
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performanceExtremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
  • Raised well rims for effective sealing, also reduces risk of cross-contaminationBatch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)