Skip to Content
Merck
All Photos(1)

Documents

EP0030122151

Eppendorf® Conical Tubes

sterile, 15 mL, colorless, pkg of 500 tubes (10 bags of 50 tubes)

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.22

material

conical bottom polypropylene

sterility

sterile

feature

with screw cap

packaging

pkg of 500 tubes (10 bags of 50 tubes)

manufacturer/tradename

Eppendorf® 0030122151

measuring range

15 mL

parameter

19500 × g max. RCF

D

17 mm

color

colorless

Looking for similar products? Visit Product Comparison Guide

General description

  • Eppendorf® conical tubes are manufactured from high-quality chemically resistant polypropylene (PP).
  • Ideal for most cell culture and cell biology applications.
  • High g-Safe centrifugation stability, and optimal sample and pellet visibility.
  • Ergonomic design - one-handed operation for easy opening/closing.
  • Free of detectable slip agents, plasticizers, and biocides ensure optimal sample integrity.
  • Pyrogen-, DNase-, RNase-, human and bacterial DNA-free for convenient and safe cell culture applications.
  • Cap with flattened and grooved sides minimizes contamination and evaporation risks.
  • A large writing area on the tube facilitates accurate labeling.
  • Compatible with centrifuge rotors, mixers, and shakers.

Application

  • Sample preparation in microbiology and molecular biology labs
  • Large volume storage of samples and solutions.
  • Preparation of mastermixes and buffers.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service