Skip to Content
Merck
All Photos(1)

Documents

EP0030108302

Eppendorf® LoBind microcentrifuge tubes

protein, 5.0 mL, colorless, pkg of 100 tubes (2 bags of 50 tubes)

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.22

sterility

non-sterile

feature

protein

packaging

pkg of 100 tubes (2 bags of 50 tubes)

manufacturer/tradename

Eppendorf® 0030108302

measuring range

5.0 mL

color

colorless

Looking for similar products? Visit Product Comparison Guide

General description

  • Eppendorf® Protein LoBind microcentrifuge tubes are made of high-quality chemically resistant polypropylene (PP).
  • LoBind material ensures minimized sample-to–surface binding for optimal product recovery.
  • PCR clean purity grade - Free from detectable human DNA, DNase, RNase, and PCR inhibitors.
  • Silicone-free tubes to minimize the risk of sample interference.
  • Features an ergonomic lid to prevent evaporation.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service