Skip to Content
Merck
All Photos(1)

Documents

EHU093191

Sigma-Aldrich

MISSION® esiRNA

targeting human CSNK1A1, CTB-89H12.4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTGGTGGAGAAATTGTGCATATGCCAATTTTTTGTTAAAACCTTTTGTTTTGAACTATACTGCTTTGAGATCTCATTTCAGAAGAACGGCATGAACAGTCTTCAGCCACAGTTGTGATGGTTGTTAAATGCTCACAATTGTGCATTCTTAGGGTTTTTCCATCCCTGGGGTTTGCAAGTTGTTCACTTAAAACATTCTTAAAATGGTTGGCTTCTTGTCTGCAAGCCAGCTGATATGGTAGCAACCAAAGATTCCAGTGTTTGAGCATATGAAAGACTCTGCCTGCTTAATTGTGCTAGAAATAACAGCATCTAAAGTGAAGACTTAAGAAAAACTTAGTGACTACTAGATTATCCTTAGGACTCTGCATTAACTCTATAATGTTCTTGGTATTAAAAAAAAAGCATATTTGTCACAGAAATTT

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luke J Fulcher et al.
EMBO reports, 20(9), e47495-e47495 (2019-07-25)
The concerted action of many protein kinases helps orchestrate the error-free progression through mitosis of mammalian cells. The roles and regulation of some prominent mitotic kinases, such as cyclin-dependent kinases, are well established. However, these and other known mitotic kinases
Xia Liu et al.
Oncogene, 39(1), 176-186 (2019-08-30)
Somatic missense mutations of the CSNK1A1 gene encoding casein kinase 1 alpha (CK1α) occur in a subset of myelodysplastic syndrome (MDS) with del(5q) karyotype. The chromosomal deletion causes CSNK1A1 haplo-insufficiency. CK1α mutations have also been observed in a variety of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service