Skip to Content
Merck
All Photos(1)

Key Documents

EHU065471

Sigma-Aldrich

MISSION® esiRNA

targeting human DOPEY1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCACGTTGCAGTCACAACTTCTTAAGGTGCTTCAGAGGCTGATTGTTCTAGAACACAGAGTAATGACTATTCCTGAAGAGAATGAAACAGGTTTTGATTTTGTTGTATCTGACTTAGAACACATCAGTCCCCATCAACCCATGACTTCTCTTCAGTATTTGCATGCTCAGCCAATCACATGTCAAGGCATGTTCCTCTGTGCAGTGATACGAGCTTTGCATCAGCACTGTGCATGTAAGATGCACCCACAATGGATTGGTTTAATCACATCTACTCTGCCTTACATGGGAAAAGTTCTGCAGAGAGTGGTTGTTTCTGTGACACTACAACTGTGCAGAAATTTAGATAATCTAATTCAGCAGTACAAATACGAAACAGGATTATCTGATAGTAGGCCTCTGTGGATGGCATCAAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ian J McGough et al.
Nature communications, 9(1), 3737-3737 (2018-09-15)
Wntless transports Wnt morphogens to the cell surface and is required for Wnt secretion and morphogenic gradients formation. Recycling of endocytosed Wntless requires the sorting nexin-3 (SNX3)-retromer-dependent endosome-to-Golgi transport pathway. Here we demonstrate the essential role of SNX3-retromer assembly for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service