Skip to Content
Merck
All Photos(1)

Key Documents

EHU058451

Sigma-Aldrich

MISSION® esiRNA

targeting human ANXA3

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€194.00
50 μG
€343.00

€194.00


Check Cart for Availability


Select a Size

Change View
20 μG
€194.00
50 μG
€343.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€194.00


Check Cart for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTTGGACACCGAGGAACAGTAAGAGATTATCCAGACTTTAGCCCATCAGTGGATGCTGAAGCTATTCAGAAAGCAATCAGAGGAATTGGAACTGATGAGAAAATGCTCATCAGCATTCTGACTGAGAGGTCAAATGCACAGCGGCAGCTGATTGTTAAGGAATATCAAGCAGCATATGGAAAGGAGCTGAAAGATGACTTGAAGGGTGATCTCTCTGGCCACTTTGAGCATCTCATGGTGGCCCTAGTGACTCCACCAGCAGTCTTTGATGCAAAGCAGCTAAAGAAATCCATGAAGGGCGCGGGAACAAACGAAGATGCCTTGATTGAAATCTTAACTACCAGGACAAGCAGGCAAATGAAGGATATCTCTCAAGCCTATTATACAGTATACAAGAAGAGTCTTGGAGATGACATTAGTTCCGAAACATCTGGTGACTTCCGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruisi Xu et al.
Journal of cellular biochemistry, 120(9), 14585-14593 (2019-04-19)
Colorectal cancer (CRC) is a common disease with high mortality and morbidity. Annexin A3 (ANXA3) belongs to the structurally homologous family of Ca2+ and phospholipid-binding proteins. This study aimed to investigate the effects and potential mechanisms of ANXA3 on oxaliplatin
S Y Yu et al.
Neoplasma, 61(3), 257-264 (2014-05-16)
Annexin A3 participates in various biological processes, including tumorigenesis, drug resistance, and metastasis. The aim of this study was to investigate the expression of Annexin A3 in gastric cancer and its relationship with cell differentiation, migration, and invasion of gastric
Stryder M Meadows et al.
PloS one, 10(7), e0132580-e0132580 (2015-07-17)
Annexins are a large family of calcium binding proteins that associate with cell membrane phospholipids and are involved in various cellular processes including endocytosis, exocytosis and membrane-cytoskeletal organization. Despite studies on numerous Annexin proteins, the function of Annexin A3 (Anxa3)
Qiu-Zhong Pan et al.
Molecular carcinogenesis, 54(8), 598-607 (2014-01-01)
Annexin A3 (ANXA3) has been found to play important roles in cancer progression, metastasis, and drug resistance; however, its role in hepatocellular carcinoma (HCC) remains unknown. In this study, we investigated the expression level, clinical significance and biologic function of
Min Jung Lee et al.
Metabolism: clinical and experimental, 64(9), 1134-1145 (2015-06-09)
Autophagy has emerged as a potentially important factor in the pathogenesis of atherosclerosis. Dehydroepiandrosterone (DHEA) is an adrenal steroid of great recent interest due to its anti-aging and anti-atherogenic effects; however, little is known about its role in autophagy and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service