Skip to Content
Merck
All Photos(1)

Key Documents

EHU008151

Sigma-Aldrich

MISSION® esiRNA

targeting human FABP3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGCCTAGCCCAGCATCACTATGGTGGACGCTTTCCTGGGCACCTGGAAGCTAGTGGACAGCAAGAATTTCGATGACTACATGAAGTCACTCGGTGTGGGTTTTGCTACCAGGCAGGTGGCCAGCATGACCAAGCCTACCACAATCATCGAAAAGAATGGGGACATTCTCACCCTAAAAACACACAGCACCTTCAAGAACACAGAGATCAGCTTTAAGTTGGGGGTGGAGTTCGATGAGACAACAGCAGATGACAGGAAGGTCAAGTCCATTGTGACACTGGATGGAGGGAAACTTGTTCACCTGCAGAAATGGGACGGGCAAGAGACCACACTTGTGCGGGAGCTAATTGATGGAAAACTCATCCTGACACTCACCCACGGCACTGCAGTTTGCACTCGCACTTATGAGAAAGAGGCATGACCTGACTGCACTGT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seung-Min Lee et al.
Nature communications, 11(1), 5661-5661 (2020-11-11)
Sarcopenia is characterized by decreased skeletal muscle mass and function with age. Aged muscles have altered lipid compositions; however, the role and regulation of lipids are unknown. Here we report that FABP3 is upregulated in aged skeletal muscles, disrupting homeostasis
Vadim Le Joncour et al.
EMBO molecular medicine, 11(6) (2019-05-10)
The current clinical care of glioblastomas leaves behind invasive, radio- and chemo-resistant cells. We recently identified mammary-derived growth inhibitor (MDGI/FABP3) as a biomarker for invasive gliomas. Here, we demonstrate a novel function for MDGI in the maintenance of lysosomal membrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service