Skip to Content
Merck
All Photos(1)

Key Documents

EHU002301

Sigma-Aldrich

MISSION® esiRNA

targeting human ITGAV

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€194.00
50 μG
€343.00

€194.00


Available to ship on03 April 2025Details



Select a Size

Change View
20 μG
€194.00
50 μG
€343.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€194.00


Available to ship on03 April 2025Details


description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATTCCACTGCAGGCTGATTTCATCGGGGTTGTCCGAAACAATGAAGCCTTAGCAAGACTTTCCTGTGCATTTAAGACAGAAAACCAAACTCGCCAGGTGGTATGTGACCTTGGAAACCCAATGAAGGCTGGAACTCAACTCTTAGCTGGTCTTCGTTTCAGTGTGCACCAGCAGTCAGAGATGGATACTTCTGTGAAATTTGACTTACAAATCCAAAGCTCAAATCTATTTGACAAAGTAAGCCCAGTTGTATCTCACAAAGTTGATCTTGCTGTTTTAGCTGCAGTTGAGATAAGAGGAGTCTCGAGTCCTGATCATGTCTTTCTTCCGATTCCAAACTGGGAGCACAAGGAGAACCCTGAGACTGAAGAAGATGTTGGGCCAGTTGTTCAGCACATCTATGAGCTGAGAAACAATGGTCCAAGTTCATTCAGCAAGGCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Zhang et al.
Cancer letters, 459, 204-215 (2019-06-15)
Macrophage-targeted therapy offers new options for intractable pancreatic ductal adenocarcinoma (PDAC), which has a low 5-year survival rate. However, the factors regulating the biological function and phenotype of macrophages in PDAC are incompletely understood. Here, we found that CD51 was
Weikun Xiao et al.
Matrix biology : journal of the International Society for Matrix Biology, 85-86, 128-146 (2019-04-28)
Originating in the brain, glioblastoma (GBM) is a highly lethal and virtually incurable cancer, in large part because it readily develops resistance to treatments. While numerous studies have investigated mechanisms enabling GBM cells to evade chemotherapy-induced apoptosis, few have addressed
Chuyue Zhang et al.
ACS nano, 14(9), 12133-12147 (2020-08-14)
Extracellular vesicles (EVs) derived from mesenchymal stem cells (MSC-EVs) have been recognized as a promising cell-free therapy for acute kidney injury (AKI), which avoids safety concerns associated with direct cell engraftment. However, low stability and retention of MSC-EVs have limited
Huashe Wang et al.
Pathology, research and practice, 215(9), 152531-152531 (2019-07-20)
Integrin subunit alpha V (ITGAV), a member of integrin family of extracellular matrix receptors, is involved in many types of cancer. In this study, the expression levels, clinical features and prognosis of ITGAV in gastric cancer (GC) patients were investigated
Hanan S Elsarraj et al.
NPJ breast cancer, 6, 12-12 (2020-05-01)
The molecular processes by which some human ductal carcinoma in situ (DCIS) lesions advance to the more aggressive form, while others remain indolent, are largely unknown. Experiments utilizing a patient-derived (PDX) DCIS Mouse INtraDuctal (MIND) animal model combined with ChIP-exo

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service