Skip to Content
Merck
All Photos(1)

Key Documents

D4297

Sigma-Aldrich

Deoxycholic acid

BioXtra, ≥98% (HPLC)

Synonym(s):

3α,12α-Dihydroxy-5β-cholanic acid, 7-Deoxycholic acid, Desoxycholic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H40O4
CAS Number:
Molecular Weight:
392.57
Beilstein:
3219882
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

biological source

bovine bile (ex gall bladder)

description

anionic

product line

BioXtra

Assay

≥98% (HPLC)

form

powder

mol wt

micellar avg mol wt 1200-5000

aggregation number

3-12

impurities

≤0.005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

CMC

2-6

mp

171-174 °C (lit.)

cation traces

Al: ≤0.001%
Ca: ≤0.01%
Cu: ≤0.0005%
Fe: ≤0.005%
K: ≤0.005%
Mg: ≤0.005%
NH4+: ≤0.05%
Na: ≤0.02%
Pb: ≤0.001%
Zn: ≤0.0005%

HLB

16

functional group

carboxylic acid

shipped in

ambient

storage temp.

room temp

SMILES string

C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

InChI

1S/C24H40O4/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3/h14-21,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-/m1/s1

InChI key

KXGVEGMKQFWNSR-LLQZFEROSA-N

Gene Information

human ... ATIC(471)

Looking for similar products? Visit Product Comparison Guide

General description

Deoxycholic acid is a bile acid that may affect intracellular signaling and gene expression. [1]

Application

Deoxycholic acid has been used in a study to assess how physiological concentrations of ursodeoxycholic acid (UDCA) vs. deoxycholic acid (DCA) affect barrier function in mouse intestinal tissue. [2] It has also been used in a study to characterize bile formation in a mouse model with hepatic disruption of the cytochrome p450 (CYP) oxidoreductase gene. [3]

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cindy Kunne et al.
Hepatology (Baltimore, Md.), 57(4), 1509-1517 (2012-11-28)
The difference in bile salt (BS) composition between rodents and humans is mainly caused by formation of muricholate in rodents as well as by efficient rehydroxylation of deoxycholic acid. The aim of this study was to characterize bile formation in
Ying Kong et al.
The international journal of biochemistry & cell biology, 44(12), 2321-2332 (2012-08-21)
There is evidence indicating that bile acid is a promoter of colorectal cancer. Deoxycholic acid modifies apoptosis and proliferation by affecting intracellular signaling and gene expression. We are interested in revealing the relationship between deregulated miRNAs and deoxycholic acid in
Kunihiko Nishino et al.
Antimicrobial agents and chemotherapy, 54(5), 2239-2243 (2010-03-10)
NlpE, an outer membrane lipoprotein, functions during envelope stress responses in Gram-negative bacteria. In this study, we report that overproduction of NlpE increases multidrug and copper resistance through activation of the genes encoding the AcrD and MdtABC multidrug efflux pumps
J Stadler et al.
Cancer letters, 38(3), 315-320 (1988-01-01)
Rectal biopsies and fecal collections were obtained from a consecutive series of 34 outpatients prior to colonoscopy at a gastroenterology clinic. Subsequently, 14 were found to have no colonic pathology, 13 had adenomatous polyps, (3 of those had a previous
Thierry Claudel et al.
Arteriosclerosis, thrombosis, and vascular biology, 25(10), 2020-2030 (2005-07-23)
Bile acids are the end products of cholesterol metabolism. They are synthesized in the liver and secreted via bile into the intestine, where they aid in the absorption of fat-soluble vitamins and dietary fat. Subsequently, bile acids return to the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service