Skip to Content
Merck
All Photos(1)

Key Documents

D1502

Sigma-Aldrich

DL-2,3-Diaminopropionic acid hydrochloride

Synonym(s):

3-Amino-DL-alanine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH(NH2)COOH · HCl
CAS Number:
Molecular Weight:
140.57
Beilstein:
5293359
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

mp

231-233 °C (dec.)

SMILES string

Cl.NCC(N)C(O)=O

InChI

1S/C3H8N2O2.ClH/c4-1-2(5)3(6)7;/h2H,1,4-5H2,(H,6,7);1H

InChI key

SKWCZPYWFRTSDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giovanni N Roviello et al.
Amino acids, 43(6), 2537-2543 (2012-06-13)
In this work, we report a synthetic approach to a Fmoc-protected nucleoamino acid, based on L-diaminopropanoic acid, carrying the DNA nucleobase on the alpha-amino group by means of an amide bond, suitable for the solid-phase synthesis of novel nucleopeptides of
J N Kalyani et al.
Journal of bacteriology, 194(20), 5604-5612 (2012-08-21)
Diaminopropionate ammonia lyase (DAPAL) is a pyridoxal-5'phosphate (PLP)-dependent enzyme that catalyzes the conversion of diaminopropionate (DAP) to pyruvate and ammonia and plays an important role in cell metabolism. We have investigated the role of the ygeX gene of Escherichia coli
Johnson Cheung et al.
Molecular microbiology, 74(3), 594-608 (2009-09-25)
Siderophores are iron-scavenging molecules produced by many microbes. In general, they are synthesized using either non-ribosomal peptide synthetase (NRPS) or NRPS-independent siderophore (NIS) pathways. Staphylococcus aureus produces siderophores, of which the structures of staphyloferrin A and staphyloferrin B are known.
Joana R Viola et al.
Bioorganic & medicinal chemistry letters, 22(17), 5635-5638 (2012-08-04)
Here we report a novel class of peptides-d-diaminopropionic acids (Dap)-for gene delivery. These peptides have attractive properties for gene delivery, and the advantage that they can be easily manipulated in relation to their composition, abiding with tailored-design. We characterized the
Changming Zhao et al.
FEBS letters, 582(20), 3125-3131 (2008-08-12)
Zwittermicin A (ZwA) is a hybrid polyketide-non-ribosomal peptide that is thought to be biosynthesized from five proposed building blocks, including the 2,3-diaminopropionate. Candidate genes for de novo biosynthesis of 2,3-diaminopropionate, zwa5A and zwa5B, have been identified in a previous study.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service