D0559
Diethylenetriaminepentaacetic acid α,ω-bis(8-stearoylamido-3,6-dioxaoctylamide) gadolinium salt
~99% (TLC)
About This Item
Assay
~99% (TLC)
reaction suitability
reagent type: catalyst
core: gadolinium
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCCCCCCC(=O)NCCOCCOCCNC(=O)CN1CCN2CCN(CC(=O)NCCOCCOCCNC(=O)CCCCCCCCCCCCCCCCC)CC(=O)O[Gd](OC(=O)C2)OC(=O)C1
InChI
1S/C62H119N7O14.Gd/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-56(70)63-35-43-80-47-49-82-45-37-65-58(72)51-68(54-61(76)77)41-39-67(53-60(74)75)40-42-69(55-62(78)79)52-59(73)66-38-46-83-50-48-81-44-36-64-57(71)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h3-55H2,1-2H3,(H,63,70)(H,64,71)(H,65,72)(H,66,73)(H,74,75)(H,76,77)(H,78,79);/q;+3/p-3
InChI key
YCQABDWULYMBBW-UHFFFAOYSA-K
Application
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service